Copper beryllium alloy (98.1:1.9 w%), alloy C17200, foil, Thickness: 0,15 mm
Catalog # 41912.GT
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1772
- Environmentally Preferable:
- Pk Info:150×150 mm
Specifications
About this item
:
:GHS08
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD01320550