Automated stainers, Cellstain® 6
THARJC651-30
- No. of slides:30
- Description:Automated stainer with six 500 ml troughs and slide holder
- Seq:003
- Environmentally Preferable:
- No. of programs:6 (May-Grünwald, Giemsa, Wright, Pappenheim, Leishman, Gram)
- W×D×H:550×280×340 mm
- Weight:15 kg
- Pk:1
Automated stainer with six staining troughs for all staining protocols with up to six different chemicals (May-Grünwald, Giemsa, Wright, Pappenheim, Leishman, Gram staining).
- Pre-programmed with six staining programs
- Programs protected by a password
- Very low consumption of stains and reagents
- The agitation and the drain time is freely selectable
- Linear staining machine to move the slide holder forward and backward
Suitable for haematology and bacteriology staining. Due to its compact size, it can be placed under a fume hood; a compatible fume extraction desk hood for vacuuming less hazardous pollutants during staining is available.
: Supplied with six staining troughs with lid, slide holder and shortened trough for loading and unloading.