Benzalkonium chloride, Sigma-Aldrich®
: Merck
: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
:63449-41-2
:8001-54-5
:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
:MFCD00145757
:29…34 °C
:0.98 g/cm³ (20 °C)
:264-151-6
:2923
:8 III
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
:GHS09
- Environmentally Preferable:
Frequently Bought Together