Tantalum (50 ppm Nb) ≥99.98% (metals basis), powder, Puratronic® -22 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-25-7
:
Formula:Ta
MDL Number:MFCD00011252
Boiling Pt:5427
Melting Pt:2996
Density:16.6 g/cm³ (20 °C)
Molecular Weight:180.95 g/mol
Storage Temperature:Ambient
EINECS:231-135-5
ADR:4.1 III
Merck Index:12,09223
UN:3089
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together