Zirconium, powder APS 2-3 µm
Catalog # 00847.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
:
:91.22 g/mol
EINECS:231-176-9
UN:1358
:GHS02
Storage Temperature:Ambient
CAS Number:7440-67-7
Formula:Zr
MDL Number:MFCD00011303
ADR:4.1 II