Order Entry
United Kingdom
ContactUsLinkComponent
 
Mercury 99.99+%
  ACRO201420025
 :  
Mercury 99.99+%
  ACRO201420025
 :  

 

  • Pk:
    2,5 kg
  • Seq:
    4076
  • Environmentally Preferable:

 

 

  • Appearance:
    Silver white Metallic liquid
  • Assay:
    ≥99.990 %
  • Cadmium (Cd):
    ≤0.1 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.5 ppm
  • Lead (Pb):
    ≤0.1 ppm
  • Silver (Ag):
    ≤1 ppm
  • Zinc (Zn):
    ≤0.5 ppm
  • Insoluble matter:
    ≤0.002 % (in HNO3)

 

 

 :
 :200.59 g/mol
 :231-106-7
 :2809
 :GHS06
 :GHS08
 :GHS09
 :Ambient
 :7439-97-6
 :Hg
 :MFCD00011035
 :356.5
 :–38.9 °C
 :13.54 g/cm³
 :8 III
 :13,05925