Acetyl-coenzyme A
A3753.0100
:
- Pk:100 mg
- CAS No.:75520-41-1
- Source:
- Species:
- Seq:02
- Environmentally Preferable:
- Storage conditions:−20 °C
- Enzyme name:Acetyl coenzyme A
- Enzyme synonyms:Acetyl-S-CoA
- Purity:83% (min.)
- Molecular weight:827,37 g/mol
- Assay (enzymatic): min. 83%
- Assay (HPLC): approx. 95%
- Loss on drying: max. 5%