Order Entry
United Kingdom
ContactUsLinkComponent
Hydroxytyrosol Acetate
Hydroxytyrosol Acetate
Catalog # ENZOALX350404M050
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Hydroxytyrosol Acetate
Catalog # ENZOALX350404M050
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    50 mg
  • CAS No.:
    69039-02-7
  • Source:
    Synthetic
  • Species:
  • Description:
    Hydroxytyrosol acetate (antioxidant), C₁₀H₁₂O₄
  • Seq:
    02
  • Environmentally Preferable:
  • Purity:
    ≥98% (HPLC)
  • Molecular weight:
    196,2
  • Storage:
    Freely soluble in organic solvents, slightly soluble in water (10 mg/ml)
  • Appearance:
    White to off-white powder

Specifications

About this item

Hydroxytyrosol acetate is an antioxidant comparable to resveratrol. It is converted to hydroxytyrosol through hydrolysis by esterases.

  • Reaches full oxygen radical absorbance capacity (ORAC) when converted to free hydoxytyrosol
  • ORAC values of hydroxytyrosol are about 50% higher than resveratrol, 10-fold higher than L(+)-ascorbic acid and 4-fold higher than (-)-epicatechin
  • Shown to have anti-cancer and anti-viral effects
  • Antibacterial at 400 µg/ml against Streptococcus pyogenes, Staphylococcus aureus, Escherichia coli and Klebsiella pneumoniae