Order Entry
United Kingdom
ContactUsLinkComponent
p70 S6K activity kit
p70 S6K activity kit
Catalog # ENZOADIEKS470
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
p70 S6K activity kit
Catalog # ENZOADIEKS470
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    p70 S6K activity kit
  • Seq:
    02
  • Environmentally Preferable:
  • Size:
    96 wells
Specifications
About this item
The p70 S6K activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
For the measurement of p70 S6K activity in partially purified, purified, or crude enzyme preparations from any species.
Ordering information: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2