Anti-Digoxigenin, Digoxin Sheep Polyclonal Antibody
Catalog # ENZOENZABS2660100
Supplier: ENZO LIFE SCIENCES
New Product
Specifications
- Antibody type:Primary
- Antigen name:Digoxigenin, Digoxin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Isotype:
- Reactivity:E. coli
- Western blot:Yes
- Seq:02
- Environmentally Preferable:
- Form:Liquid
- Storage buffer:In PBS containing 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Digoxin
- Purification:Protein A affinity purified
- Pk:100 µG
Specifications
About this item
Recommended Applications: ELISA, WB, TUNEL, ISH (in situ hybridization)
Species reactivity: Species independent
Type: Primary
Antigen: Digoxigenin, Digoxin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: E. coli