Anti-IL7R Mouse Monoclonal Antibody (Alexa Fluor® 546) [clone: R34-34]
ENZOENZABS2500100
New Product
- Antibody type:Primary
- Antigen name:IL-7 receptor-alpha
- Antigen symbol:IL7R
- Clonality:Monoclonal
- Clone:R34-34
- Conjugation:Alexa Fluor® 546
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:
- Seq:02
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:IL-7RA|Interleukin-7 receptor α|CD127
- Storage buffer:In PBS containing 50% glycerol.
- Storage temperature:–80 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Human leukemic pre-B cells.
- Pk:100 µG
Recommended Applications: IF
Species reactivity: Human, Dog
Type: Primary
Antigen: IL7R
Clonality: Monoclonal
Clone: R34-34
Conjugation: Alexa Fluor® 546
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human