- Antibody type:Primary
- Antigen name:N-Ethylmaleimide-Sensitive Fusion Protein
- Antigen symbol:NSF
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Environmentally Preferable:
- Form:Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide.
- Purification:Protein G affinity purified
- Pk:50 µG
N-ethylmaleimide-sensitive factor (NSF) is involved in a variety of membrane fusion events and plays a prominent role in synaptic vesicle fusion. In order to interact with target membranes, NSF requires another set of soluble proteins called Soluble NSF attachment proteins (SNAPs). Together with synaptotagmin and alpha-SNAP, NSF modulates the interaction of SNAP25, VAMP, and syntaxin in an ATP-dependent manner to form a 20S complex involved in synaptic vesicle docking and fusion.
- For Western Blot
Recognizes human, mouse, rat, bovine, dog, chicken, hamster, monkey, pig, rabbit and sheep NSF. Detects a band of ~76kDa by Western blot.
Type: Primary
Antigen: N-ETHYLMALEIMIDE-SENSITIVE FUSION PROTEIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human