Biological indicators, spore threads
Supplier: CROSSTEX
Certificates
About this item
Spore threads are prepared from Geobacillus stearothermophilus. These are designed to be placed directly into a device and utilised to monitor steam sterilisation process efficacy.
Ideal for use in tubing, lumens and other devices/instruments where larger BIs and carriers cannot be utilised.
Frequently Bought Together