Benzyl alcohol 98.0-100.5% (by acidometric titration), liquid USP, NF, Ph. Eur., BP, pharma grade
Supplier: PanReac AppliChem
Certificates
About this item
Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol
CAS Number:100-51-6
:
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599
Boiling Pt:204.7 °C
Melting Pt:–15,2 °C
Density:1.043 – 1.049 (20°C)
Molecular Weight:108.14 g/mol
Flash Pt:94 °C
Merck Index:13,01126
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearence:Clear colourless liquid
- Solubility in water:Soluble in water 40 g/L at 20 °C D 20/4 1,0453
- Flash point:94 °C 0,13 hPa
- Vapor pressure:20 °C
- Assay (acidic medium):98,0 – 100,5%
- Refractive index n20/D:1,5396
- Ignition temperature:435 °C
- Dielectric constant:25 °C 13,1
- Heat evaporation:205 °C 467 KJ/Kg
- Saturation concentration:20 °C 0,56 g/m³
- Explosion limit:1,3% (V) 13% (V)
- Density:1,043 – 1,049 (20/20)
- Non-volatile matter:0,05%
- Residue on ignition (as SO₄) :0,005%
- Peroxide value:5
- Benzaldehyde (G.C.):0,15%
- Ciclohexylmethanol:0,10%
- Halogenated compounds and halides (as Cl):0,03%
- Water (H₂O) :0,2%
- Cadmium (Cd):0,5 ppm
- Lead (Pb):0,5 ppm
- Arsenic (As):1,5 ppm
- Mercury (Hg):1,5 ppm
- Cobalt (Co):5 ppm
- Vanadium (V):10 ppm
- Nickel:20 ppm
- Titanium:0,8 ppm
- Gold (Au):10 ppm
- Palladium (Pd):10 ppm
- Iridium:10 ppm
- Osmium (Os):10 ppm
- Rhodium (Rh):10 ppm
- Ruthenium (Ru):10 ppm
- Selenium (Se):10 ppm
- Silver (Ag):10 ppm
- Platinum (Pt):10 ppm
- Lithium (Li):55 ppm
- Antimony (Sb):120 ppm
- Barium (Ba):140 ppm
- Molybdenum:25 ppm
- Copper (Cu):250 ppm
- Tin (Sn):600 ppm
- Chromium (Cr):25 ppm
- Total imp. Tr < benzyl alcohol:0,04%
- Passes test:0,3%
- undefined:Residual solvents (Ph.Eur/USP)
Specification Test Results
Frequently Bought Together