Order Entry
United Kingdom
ContactUsLinkComponent
Anti-P GlycoProtein Polyclonal Antibody
Anti-P GlycoProtein Polyclonal Antibody
Catalog # BSBTRP1034
Supplier:  Boster Bio
undefined
Anti-P GlycoProtein Polyclonal Antibody
Catalog # BSBTRP1034
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    P GlycoProtein
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for 1853 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. 1853 information: Molecular Weight: 141479 MW; Subcellular Localization: Cell membrane ; Multi-pass membrane protein ; Tissue Specificity: Expressed in liver, kidney, small intestine and brain.

Type: Primary
Antigen: P GlycoProtein
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat