Anti-MPO/Myeloperoxidase Polyclonal Antibody
Catalog # BSBTPB9057
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Myeloperoxidase
- Antigen symbol:MPO
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for Myeloperoxidase/MPO detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Myeloperoxidase/MPO information: Molecular Weight: 83869 MW; Subcellular Localization: Lysosome.
Type: Primary
Antigen: MPO/Myeloperoxidase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat