(R)-3-Methylmorpholine hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:953780-78-4
:
Formula:C₅H₁₂ClNO
MDL Number:MFCD18382515
Molecular Weight:137.6100 g/mol
Melting Pt:126…128.5 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow to light brown Powder or solid
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 to ≤104.0 %
- Specific optical rotation:+1° to +3° (20°C, 589 nm) c=1 in H2O
Specification Test Results
Frequently Bought Together