Doxycycline hyclate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Doxycycline Hydrochloride Hemiethanolate Hemihydrate
CAS Number:24390-14-5
:
Formula:C₂₂H₂₄N₂O₈·HCl·0.5H₂O·0.5C₂H₆O
MDL Number:MFCD07357237
Molecular Weight:512.94 g/mol (503.94 anhy)
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Water:1.4 to 2.8 %
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together