Promethazine hydrochloride
: 10-[2-(Dimethylamino)propyl]phenothiazine Hydrochloride, N,N-Dimethyl-1-(10H-phenothiazin-10-yl)propan-2-amine hydrochloride
:58-33-3
:
:C₁₇H₂₀N₂S
:MFCD00012652
:320.88 g/mol
:227…230 °C
:3077
:9 III
:GHS07
:GHS09
:Ambient
- Environmentally Preferable:
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C, 4 hr)
- Solubility:(10% in water) Clear colorless to light yellow
- Titration with HClO4:97.5 to 101.5 % (on dry substance)
Frequently Bought Together