Bromine (bromide-bromate) solution 0.05 M Br2 (0.1N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
:
MDL Number:MFCD00143870
Density:1.01 g/cm³
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:0.0950 - 0.1050 N
Specification Test Results
Frequently Bought Together