VWR® PH 20 pH Meter, Handheld
662-2283
- Description:Handheld pH meter, pH 20
- Environmentally Preferable:
- Model:pH 20
- Compliance:IP 65 / IP 67
- Connections:BNC
- Display:3-line display with backlight
- Power supply:2 AA batteries
- Temperature compensation:−5...+150 °C (manual)
- Weight:0,13 kg (without electrode)
- W×D×H:54×28×108 mm
- pH accuracy:±0,02 pH ±1 digit
- pH calibration:Up to 2 points
- pH range:0,00 - 14,00
- pH resolution:0,01
- Pk:1
This handheld measuring device features an ergonomic design, dust and waterproof housing in accordance with IP 65/67 and an illuminated display. This compact pH meter is a perfect alternative to pH testers.
- Modern and functional ABS housing
- 3-line display with backlight
- HOLD function, min./max. function, low battery indicator
- Durable, long battery life
- BNC connection for alternating electrodes
Typical applications include: Aquariums and aquaculture, plant cultivation and agriculture, laboratories, quality assurance, service and food.
: Supplied with epoxy pH electrode, buffer capsules (2× pH 4 and 2× pH 7), 2× AA batteries and manual.