Standards for water pollution QC check, solids, VHG
This product is intended for use as a quality control check for methods related to total suspended solids (TSS), total dissolved solids (TDS), and total solids (TS).
This standard has been prepared as a concentrate and must be diluted prior to analysis. This standard is not preserved but can be stored at room temperature.
: Traceable to NIST SRM.
- Environmentally Preferable:
Frequently Bought Together