Nitric acid fuming ≥99%, AnalaR NORMAPUR®
Catalog # 85850.270
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle, plastic safety coating
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:min. 99.0 %
- Appearance:yellow clear liquid
- Chloride (Cl):max. 0.0005 %
- Heavy metals (as Pb):max. 0.0005 %
- Sulphate (SO4):max. 0.001 %
- Ag (Silver):max. 0.00001 %
- Al (Aluminium):max. 0.0001 %
- As (Arsenic):max. 0.00002 %
- Ba (Barium):max. 0.00001 %
- Be (Beryllium):max. 0.00001 %
- Bi (Bismuth):max. 0.00005 %
- Ca (Calcium):max. 0.00005 %
- Cd (Cadmium):max. 0.00001 %
- Co (Cobalt):max. 0.00001 %
- Cr (Chromium):max. 0.00005 %
- Cu (Copper):max. 0.00001 %
- Fe (Iron):max. 0.0005 %
- K (Potassium):max. 0.00005 %
- Li (Lithium):max. 0.00001 %
- Mg (Magnesium):max. 0.00001 %
- Mn (Manganese):max. 0.00001 %
- Mo (Molybdenum):max. 0.00001 %
- Na (Sodium):max. 0.0003 %
- Ni (Nickel):max. 0.00005 %
- Pb (Lead):max. 0.00001 %
- Ti (Titanium):max. 0.00005 %
- V (Vanadium):max. 0.00001 %
- Zn (Zinc):max. 0.0003 %
- Zr (Zirconium):max. 0.001 %
- residue on ignition (as SO4):max. 0.005 %
Specification Test Results
About this item
Synonyms: Nitric acid solution
:
:63.01 g/mol
EINECS:231-714-2
UN:2032
:GHS03
:GHS05
:GHS06
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349
Boiling Pt:86 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.51 g/cm³ (20 °C)
ADR:8 I
Merck Index:13,06610