Fluvastatin sodium salt
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: XU 62-320, rel-(3S,5R,E)-7-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoic acid sodium salt, Sodium rel-(3R,5S,E)-7-(3-(4-fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoate
CAS Number:93957-55-2
:93957-54-1
Formula:C24 H25FNNaO4
MDL Number:MFCD00929076
Molecular Weight:433.46 g/mol
:GHS07
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Water:≤4 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together