Graphene, nanoplatelets 2-10 nm
Catalog # ACRO453160500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Grey to black Powder (Consists of stacks of multi-layer graphene sheets)
- Additional info:Tensile strength: 5 Gpa
- Particle size:2 to 10 nm (Thickness)
- Specific surface area:20 to 40 m²/g
- Conductivity:80000 S/m
Specification Test Results
About this item
:
Storage Temperature:Ambient
CAS Number:1034343-98-0
:7782-42-5
MDL Number:MFCD00144065
Melting Pt:3650 °C