Metanil yellow
: 3-(4-Anilinophenylazo)benzenesulphonic acid sodium salt, Acid Yellow 36
:587-98-4
:
:C₁₈H₁₄N₃NaO₃S
:MFCD00007486
:375.38 g/mol
:GHS08
:GHS09
:Ambient
- Environmentally Preferable:
- Appearance:Golden to orange Powder
- Infrared spectrum:Conforms
- Dye content:≥88 %
- UV Lambda max:413 to 419 nm (in methanol)
- Specific absorbance A (1%/1cm):≥400
- Visual transition interval:from pH 1.2 (red) to 2.3 (yellow)
Frequently Bought Together