Human recombinant vitronectin (from cells)
Catalog # PRSI91-362
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Environmentally Preferable:
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Vitronectin|Serum-Spreading Factor|S-Protein|VTN|V75|VN
- Protein/peptide name:Vitronectin
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:53.35 kD
- Sequence:Asp20-Leu478
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Human Vitronectin/VTN is a cell adhesion and spreading factor. It can be found in the blood and the extracellular matrix (ECM). Vitronectin interacts with glycosaminoglycans and proteoglycans. The multimeric Vitronectin can efficiently bind to and incorporate into the ECM; Vitronectin can support cell adhesion through binding to various integrins and other proteoglycans. Vitronectin can be recognized by certain members of the integrin family and serves as a cell-to-substrate adhesion molecular. It can as a inhibitor of the membrane-damaging effect of the terminal cytolytic complement pathway. Vitronectin contains an endogenous cleavage site, plus cleavage sites for elastase, thrombin, and plasmin.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag