Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Environmentally Preferable:
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus PD-L1, His Tag at 10 ug/mL (100 ul/well) can bind Cynomolgus PD-1, Fc Tag with a linear range of 0.6-10 ug/mL.
- Protein synonyms:B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
- Protein/peptide name:PDL14
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:27.1 kDa
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot
Specifications
About this item
Programmed cell death 1 ligand 1 (PDL1) is also known as B7-H, B7H1, MGC142294, MGC142296, PD-L1, PDCD1L1 and PDCD1LG1,which is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses.PDL1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. This protein is broadly expressed in the majority of peripheral tissues as well as hematopoietic cells. Interaction between PDL1 and its receptors belonging to the CD28 family of molecules provide both stimulatory and inhibitory signals in regulating T cell activation and tolerance. PDL1 may inhibit ongoing T-cell responses by inducing apoptosis and arresting cell-cycle progression.