Human MAP-1 peptide (15 aa in the center)
PRSI3005P
:
- Pk:50 µG
- Source:
- Species:Human
- Environmentally Preferable:
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa in the center
- Protein/peptide name:MAP-1 Peptide (15 aa in the center)
- Tested applications:Blocking
MAP1 peptide is used for blocking the activity of the MAP-1 antibody.