Anti-PPAPDC2 Rabbit Polyclonal Antibody
Catalog # PRSI5517
Supplier: ProSci Inc.
Specifications
- Antigen name:Phosphatidic Acid Phosphatase Type 2 Domain Containing 2
- Antigen symbol:PPAPDC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:Phosphatidic acid phosphatase type 2 domain-containing protein 2|PSDP|Presqualene diphosphate phosphatase|PDP1|bA6J24.6|PPAPDC2|PPAP2 domain-containing protein 2
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:PPAPDC2 antibody was raised against a 13 amino acid synthetic peptide from near the carboxy terminus human PPAPDC2.
- Tested applications:E, WB, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type:
Antigen: PPAPDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat