- Antigen name:C-Reactive Protein
- Antigen symbol:CRP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:CRP1|MGC149895|C-reactive protein|pentraxin 1|CRP|D1S181E|RP11-419N10.4|CSRP|CYRP|MGC88244|PTX1
- Concentration:1 mg/mL
- Immunogen:Full Length
- Size:50 µg
- Pk:50 µG
Cysteine-rich protein 2; Cystein-rich intestinal protein. C-Reactive Protein is a 208-amino acid protein containing 2 LIM domains and shares 93% amino acid sequence identity with its rat homolog, called Esp1 or Crp2. Northern blot analysis showed widespread tissue expression of the 1.3-kb C-Reactive Protein mRNA, with the highest level of expression found in heart. In testis, a second 1.7-kb mRNA was also detected.
CRP antibody can be used for the detection of CRP by Western blot, ELISA, may also work for IHC and ICC.
Antigen affinity-purified
Phosphate-Buffered Saline. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: