Specifications
- Antigen name:Interleukin 6
- Antigen symbol:IL6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Neutralisation:Yes
- Form:Lyophilized
- Antigen synonyms:HSF|HGF|IFNB2|interferon beta 2|BSF2|IL-6
- Immunogen:Produced from sera of goats pre-immunized with highly pure (>98%) recombinant Rat IL-6.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Interleukin 6 (IL-6) is a multifunctional 24 kDa protein originally discovered in the medium of RNA stimulated fibroblastoid cells. It is upregulated by IL1, TNF, PDGF, IFN beta, TNF alpha, NGF, IL17 and downregulated by glucocorticoids IL4, TGF beta. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be as important as IL1 and TNF alpha in regulating the acute phase response. IL-6 has also been implicated in regulating adipose mass.
Anti-Rat IL-6 specific antibody was purified by affinity chromatography employing immobilized Rat IL-6 matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat