Anti-NT3 Chicken Polyclonal Antibody
PRSIXG-6103
- Antigen name:Neurotrophin 3
- Antigen symbol:NT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:NGF-2|HDNF|Ntf3|Neurotrophin-3|Nerve growth factor 2|Ntf-3|Neurotrophic factor
- Immunogen:NT3 Mixed Peptides
- Size:500 µg
- Pk:500 µG
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse