Anti-GYS1 Rabbit Polyclonal Antibody
PRSI79-716
- Antigen name:Glycogen Synthase 1
- Antigen symbol:GYS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Glycogen synthase 1 phospho Ser645|EC 2.4.1.11|UDP glucose glycogen glucosyltransferase|GYS 1|GYS1_HUMAN|p-Glycogen synthase 1 S645|Glycogen synthase1|GSY|Starchsynthase muscle|GYS|Glycogen synthase 1|Glycogen [starch] synthase|GYS1|muscl.|Glycogen synthase 1 phospho S645|Glycogen synthase 1 muscle
- Concentration:1 mg/mL
- Immunogen:Glycogen Synthase antibody was raised against a peptide sequence around aa. 644~648 (P-S-P-S-L) derived from Human Glycogen Synthase .
- Size:100 µg
- Pk:100 µG
Transfers the glycosyl residue from UDP-Glc to the non-reducing end of alpha-1, 4-glucan.
Western Blot: 1:500
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GYS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat