Specifications
- Antigen name:Stathmin 1
- Antigen symbol:STMN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Op18|SMN|C1orf215|PP19|LAP18|OP18|Stathmin|Lag|PR22|Leukemia-associated phosphoprotein p18|PP17
- Concentration:1 mg/mL
- Immunogen:Stathmin 1 (Ab-16) antibody was raised against a peptide sequence around aa.14~18 (R-A-S-G-Q) derived from Human Stathmin 1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Involved in the regulation of the microtubule (MT) filament system by destabilizing microtubules. Prevents assembly and promotes disassembly of microtubules. Phosphorylation at Ser-16 may be required for axon formation during neurogenesis. Involved in the control of the learned and innate fear
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: STMN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat