Anti-TSC22D3 Mouse Monoclonal Antibody
PRSI51-450
- Antibody type:Tag
- Antigen name:TSC22 Domain Family Member 3
- Antigen symbol:TSC22D3
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:DSIPI|Protein DIP|GILZ|DSIP-immunoreactive peptide|hDIP|TSC-22R|DIP|TSC22 domain family protein 3|TSC22D3
- Immunogen:TSC22D3 (NP_932174, 1 a.a. ~ 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:50 µg
- Pk:50 µG
TSC22D3 antibody can be used in Western Blot starting at 1 - 2 μg/mL, and immunohistochemistry starting at 5 μg/mL.
Protein A Column
PBS, pH 7.2
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Tag
Antigen: TSC22D3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human