- Antigen name:Transient Receptor Potential Cation Channel Subfamily V member 2
- Antigen symbol:TRPV2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Pig,Horse,Monkey,Gorilla,Human
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:S|osm-9-like TRP channel 2|OTRPC2|Transient receptor potential cation channel subfamily V member 2|TrpV2|Vanilloid receptor-like protein 1|VRL-1
- Immunogen:TRPV2 antibody was raised against a peptide located in the N-Terminal domain of TRPV2 (Human).
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
Transient receptor potential vanilloid (TRPV) channels are widely expressed in both sensory and nonsensory cells. TRPV2, a member of transient receptor potential ion channels, responds to high-threshold noxious heat (with a threshold of approximately 52 degrees C), but neither to capsaicin nor to proton. TRPV2 transcripts are not restricted to the sensory nervous system, indicating that this channel may be activated by stimuli other than heat. It has been proposed that responses to noxious heat involve these related, but distinct, ion-channel subtypes that together detect a range of stimulus intensities.
TRPV2 antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200 - 1:400, and immunofluorescence starting at 1:200 - 1:1000.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRPV2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Horse, Human, Monkey, Pig