Anti-APOB Goat Polyclonal Antibody
Catalog # PRSI46-802
Supplier: ProSci Inc.
Specifications
- Antigen name:Apolipoprotein B
- Antigen symbol:APOB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:apoB-100|apo B-100|FLDB|Apolipoprotein B-100|Apolipoprotein B46|apoB-48|LDLCQ4
- Concentration:500 ug/mL
- Immunogen:Apolipoprotein B antibody was raised against a 12 amino acid synthetic peptide near the internal region of Apolipoprotein B.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Bands corresponding to the exopected sizes of ApoB100 and ApoB48 were detected in human serum samples from five different individuals. Recommended concentration: 0.3-1 μg/mL.
Apolipoprotein B antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Apolipoprotein B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: APOB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human