Anti-APOL5 Goat Polyclonal Antibody
PRSI46-747
- Antigen name:Apolipoprotein L5
- Antigen symbol:APOL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:APOLV|APOL-V
- Concentration:500 ug/mL
- Immunogen:ApoL5 antibody was raised against a 14 amino acid synthetic peptide near the C-Terminus of ApoL5.
- Size:100 µg
- Pk:100 µG
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 48 kDa band observed in human frontal cortex lysates (calculated MW of 47.0 kDa according to NP_085145.1). Recommended concentration: 1-3 μg/mL.
ApoL5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ApoL5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: APOL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human