- Antigen name:Patatin-like Phospholipase Domain Containing 3
- Antigen symbol:PNPLA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Adiponutrin|C22orf20|PNPLA3|iPLA(2)epsilon|Adpn
- Concentration:500 ug/mL
- Immunogen:PNPLA3 antibody was raised against a 12 amino acid synthetic peptide near the internal region of PNPLA3.
- Size:100 µg
- Pk:100 µG
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 45 kDa band observed in human heart lysates (calculated MW of 42.6 kDa according to NP_473429.2). Recommended concentration: 0.03-0.1 μg/mL. Approximately 45 kDa band observed in mouse heart lysates (calculated MW of 42.6 kDa according to human NP_473429.2 and to mouse NP_473429.2). Recommended concentration: 0.05-0.2 μg/mL. Immunohistochemistry: In paraffin embedded human Skin shows membranous staining of basak kayer cells. Recommended concentration, 4-6 μg/mL.
PNPLA3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
PNPLA3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PNPLA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse