Anti-ARSA Goat Polyclonal Antibody
Catalog # PRSI46-627
Supplier: ProSci Inc.
Specifications
- Antigen name:Arylsulfatase A
- Antigen symbol:ARSA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Arylsulfatase A|DRIL3|ARSA|DRIL1|Cerebroside-sulfatase|MLD|ASA|BRIGHT|E2FBP1
- Concentration:500 ug/mL
- Immunogen:Arylsulfatase A antibody was raised against a 12 amino acid synthetic peptide near the internal region of Arylsulfatase A.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 55 kDa band observed in mouse testis lysates (calculated MW of 53.6 kDa according to human NP_000478.2 and 53.7 kDa according to mouse NP_033843.2). Recommended concentration: 0
Arylsulfatase A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Arylsulfatase A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ARSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse