Anti-WDFY1 Goat Polyclonal Antibody
Catalog # PRSI46-592
Supplier: ProSci Inc.
Specifications
- Antigen name:WD Repeat And FYVE Domain Containing 1
- Antigen symbol:WDFY1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:WDF1|ZFYVE17|FENS-1
- Concentration:500 ug/mL
- Immunogen:WDFY1 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of WDFY1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
WDFY1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
WDFY1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: WDFY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human