Anti-VDAC2 Goat Polyclonal Antibody
PRSI46-570
- Antigen name:Voltage-dependent Anion Channel 2
- Antigen symbol:VDAC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Concentration:500 ug/mL
- Immunogen:VDAC2 antibody was raised against an 11 amino acid synthetic peptide near the internal region of VDAC2.
- Size:100 µg
- Pk:100 µG
VDAC2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
VDAC2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: VDAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human