Order Entry
United Kingdom
ContactUsLinkComponent
Anti-TIA1 Goat Polyclonal Antibody
Anti-TIA1 Goat Polyclonal Antibody
Catalog # PRSI46-492
Supplier:  ProSci Inc.
undefined
Anti-TIA1 Goat Polyclonal Antibody
Catalog # PRSI46-492
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    TIA1 cytotoxic granule-associated RNA binding protein
  • Antigen symbol:
    TIA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    TIA-1|WDM
  • Concentration:
    500 ug/mL
  • Immunogen:
    TIA1 antibody was raised against an 11 amino acid synthetic peptide near the internal region of TIA1.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

TIA1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TIA1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TIA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human