Anti-PRICKLE3 Goat Polyclonal Antibody
Catalog # PRSI46-221
Supplier: ProSci Inc.
Specifications
- Antigen name:Prickle Homolog 3
- Antigen symbol:PRICKLE3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:LMO6
- Concentration:500 ug/mL
- Immunogen:PRICKLE3 antibody was raised against an 11 amino acid synthetic peptide near the internal region of PRICKLE3.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
PRICKLE3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
PRICKLE3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PRICKLE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human