Anti-PLIN1 Goat Polyclonal Antibody
Catalog # PRSI46-167
Supplier: ProSci Inc.
Specifications
- Antigen name:Perilipin 1
- Antigen symbol:PLIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:PLIN|perilipin|PERI|FPLD4|lipid droplet-associated protein
- Concentration:500 ug/mL
- Immunogen:Perilipin antibody was raised against a 12 amino acid synthetic peptide near the internal region (near the C-Terminus) of Perilipin.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Perilipin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Perilipin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PLIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human