Anti-NOTCH2 Goat Polyclonal Antibody
PRSI45-977
- Antigen name:Notch 2
- Antigen symbol:NOTCH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:AGS2|hN2|HJCYS
- Concentration:500 ug/mL
- Immunogen:NOTCH2 antibody was raised against a 14 amino acid synthetic peptide near the internal region of NOTCH2.
- Size:100 µg
- Pk:100 µG
NOTCH2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
NOTCH2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NOTCH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0