Anti-NET1 Goat Polyclonal Antibody
PRSI45-947
- Antigen name:Neuroepithelial Cell Transforming 1
- Antigen symbol:NET1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:ARHGEF8|NET1A|neuroepithelial cell transforming gene 1|NET1|Rho guanine nucleotide exchange factor (GEF) 8|p65 Net1 proto-oncogene protein|guanine nucleotide regulatory protein (oncogene)|small GTP-binding prot|neuroepithelioma transforming gene 1
- Concentration:500 ug/mL
- Immunogen:NET1 antibody was raised against a 14 amino acid synthetic peptide near the C-Terminus of NET1.
- Size:100 µg
- Pk:100 µG
NET1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
NET1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NET1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human