Order Entry
United Kingdom
ContactUsLinkComponent
Anti-LIN7B Goat Polyclonal Antibody
Anti-LIN7B Goat Polyclonal Antibody
  PRSI45-832
undefined
Anti-LIN7B Goat Polyclonal Antibody
  PRSI45-832

 

  • Antigen name:
    Lin-7 homolog B
  • Antigen symbol:
    LIN7B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    MALS-2|LIN-7B|VELI2|MALS2
  • Concentration:
    500 ug/mL
  • Immunogen:
    LIN7B antibody was raised against a 14 amino acid synthetic peptide near the N-Terminus of LIN7B.
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

LIN7B antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

LIN7B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LIN7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat