Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-CYTH4 Goat Polyclonal Antibody
Catalog # PRSI45-453
Supplier:  ProSci Inc.
undefined
Anti-CYTH4 Goat Polyclonal Antibody
Catalog # PRSI45-453
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Cytohesin 4
  • Antigen symbol:
    CYTH4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    CYT4|PSCD4|DJ63G5.1
  • Concentration:
    500 ug/mL
  • Immunogen:
    Cytohesin 4 antibody was raised against a 13 amino acid synthetic peptide near the N-Terminus of Cytohesin 4.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Cytohesin 4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Cytohesin 4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CYTH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: